Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human CCR6 Alexa Fluor™ 405-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FAB195RV100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB195RV100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB195RV100 Supplier R&D Systems Supplier No. FAB195RV100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

CCR6 Monoclonal specifically detects CCR6 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen CCR6
Applications Flow Cytometry
Classification Monoclonal
Clone 53103R
Conjugate Alexa Fluor 405
Dilution Flow Cytometry 0.25-1 μg/10^6 cells
Formulation Supplied 0.2 mg/ml in a saline solution containing BSA and Sodium Azide.
Gene Accession No. P51684
Gene Alias BN-1, CC-CKR-6, CCR6, CCR-6, CD196, CD196 antigen, chemokine (C-C motif) receptor 6, CKRL3, CKR-L3, CKR-L3G-protein coupled receptor 29, CKRL3LARC receptor, CMKBR6, CMKBR6STRL22, DRY6, GPR29, GPR29C-C CKR-6, GPRCY4, GPR-CY4, GPRCY4DRY-6, lymphocyte, 22, STRL22
Host Species Mouse
Immunogen NS0 mouse myeloma cell line transfected with human CCR6 Met1-Met374 Accession # P51684
Purification Method Protein A or G purified from cell culture supernatant
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 1235
Test Specificity Detects human CCR6. Stains human CCR6 transfectants but not the parental cell line in flow cytometry.
Target Species Human
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Form Supernatant
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.