Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human CCR6 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB1951100
This item is not returnable.
View return policy
Description
CCR6 Monoclonal specifically detects CCR6 in Human samples. It is validated for Flow Cytometry, CyTOF-ready.Specifications
CCR6 | |
Monoclonal | |
Unconjugated | |
Lyophilized from a 0.2 µm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 µm filtered solution in PBS. | |
BN-1, CC-CKR-6, CCR6, CCR-6, CD196, CD196 antigen, chemokine (C-C motif) receptor 6, CKRL3, CKR-L3, CKR-L3G-protein coupled receptor 29, CKRL3LARC receptor, CMKBR6, CMKBR6STRL22, DRY6, GPR29, GPR29C-C CKR-6, GPRCY4, GPR-CY4, GPRCY4DRY-6, lymphocyte, 22, STRL22 | |
NS0 mouse myeloma cell line transfected with human CCR6, Met1-Met374, Accession # P51684 | |
100 μg | |
Primary | |
Detects human CCR6 in direct ELISAs. | |
Human | |
Purified |
Flow Cytometry, CyTOF | |
1011306 | |
Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready | |
P51684 | |
Mouse | |
Protein A or G purified from ascites | |
RUO | |
1235 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction