Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human CCR7 Alexa Fluor™ 647-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems FAB197R025
This item is not returnable.
View return policy
Description
CCR7 Monoclonal specifically detects CCR7 in Human samples. It is validated for Flow Cytometry.Specifications
CCR7 | |
Monoclonal | |
Alexa Fluor 647 | |
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
BLR2, BLR2C-C chemokine receptor type 7, C-C CKR-7, CC-CKR-7, CCR7, CCR-7, CD197, CD197 antigen, CDw197, CDw197CC chemokine receptor 7, chemokine (C-C motif) receptor 7, CMKBR7, CMKBR7chemokine (C-C) receptor 7, EBI1, EBI1EBV-induced G protein-coupled receptor 1, EBV-induced G-protein coupled receptor 1, Epstein-Barr virus induced gene 1, Epstein-Barr virus induced G-protein coupled receptor, Epstein-Barr virus-induced G-protein coupled receptor 1, EVI1, lymphocyte-specific G protein-coupled peptide receptor, MIP-3 beta receptor | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
1236 | |
Human | |
Supernatant |
Flow Cytometry | |
150503 | |
Flow Cytometry 5 uL/10^6 cells | |
AAA58615 | |
CCR7 | |
Human CCR7 transfectants Met1-Pro378 Accession # AAA58615 | |
25 Tests | |
Primary | |
Detects human CCR7. | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction