Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human CCR7 Alexa Fluor™ 700-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 3 publications
$139.50 - $456.00
Specifications
Antigen | CCR7 |
---|---|
Clone | 150503 |
Dilution | Flow Cytometry 5 uL/10^6 cells |
Applications | Flow Cytometry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
FAB197N025
![]() |
R&D Systems
FAB197N025 |
25 Tests |
Each for $139.50
|
|
|||||
FAB197N100
![]() |
R&D Systems
FAB197N100 |
100 Tests |
Each for $456.00
|
|
|||||
Description
CCR7 Monoclonal specifically detects CCR7 in Human samples. It is validated for Flow Cytometry.Specifications
CCR7 | |
Flow Cytometry 5 uL/10^6 cells | |
Monoclonal | |
Purified | |
Human | |
BLR2, BLR2C-C chemokine receptor type 7, C-C CKR-7, CC-CKR-7, CCR7, CCR-7, CD197, CD197 antigen, CDw197, CDw197CC chemokine receptor 7, chemokine (C-C motif) receptor 7, CMKBR7, CMKBR7chemokine (C-C) receptor 7, EBI1, EBI1EBV-induced G protein-coupled receptor 1, EBV-induced G-protein coupled receptor 1, Epstein-Barr virus induced gene 1, Epstein-Barr virus induced G-protein coupled receptor, Epstein-Barr virus-induced G-protein coupled receptor 1, EVI1, lymphocyte-specific G protein-coupled peptide receptor, MIP-3 beta receptor | |
CCR7 | |
IgG2a | |
Protein A or G purified from hybridoma culture supernatant | |
Detects human CCR7. |
150503 | |
Flow Cytometry | |
Alexa Fluor 700 | |
Mouse | |
AAA58615 | |
1236 | |
Human CCR7 transfectants Met1-Pro378 Accession # AAA58615 | |
Primary | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title