Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human CCR7 APC-conjugated Antibody, R&D Systems™
GSA_VAGREENER_CHOICE

Catalog No. FAB197A100
Change view
Click to view available options
Quantity:
100 Tests
25 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB197A100 100 Tests
FAB197A025 25 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. FAB197A100 Supplier R&D Systems Supplier No. FAB197A100
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody has been used in 18 publications

CCR7 Monoclonal specifically detects CCR7 in Human samples. It is validated for Flow Cytometry.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Antigen CCR7
Applications Flow Cytometry
Classification Monoclonal
Clone 150503
Conjugate APC
Dilution Flow Cytometry 10 uL/10^6 cells
Gene Accession No. AAA58615
Gene Alias BLR2, BLR2C-C chemokine receptor type 7, C-C CKR-7, CC-CKR-7, CCR7, CCR-7, CD197, CD197 antigen, CDw197, CDw197CC chemokine receptor 7, chemokine (C-C motif) receptor 7, CMKBR7, CMKBR7chemokine (C-C) receptor 7, EBI1, EBI1EBV-induced G protein-coupled receptor 1, EBV-induced G-protein coupled receptor 1, Epstein-Barr virus induced gene 1, Epstein-Barr virus induced G-protein coupled receptor, Epstein-Barr virus-induced G-protein coupled receptor 1, EVI1, lymphocyte-specific G protein-coupled peptide receptor, MIP-3 beta receptor
Gene Symbols CCR7
Host Species Mouse
Immunogen Human CCR7 transfectants Met1-Pro378 Accession # AAA58615
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 Tests
Primary or Secondary Primary
Gene ID (Entrez) 1236
Test Specificity Detects human CCR7.
Target Species Human
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.