Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human CCR9 APC-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FAB1791A025
Change view
Click to view available options
Quantity:
100 Tests
25 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB1791A025 25 Tests
FAB1791A100 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. FAB1791A025 Supplier R&D Systems Supplier No. FAB1791A025
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody has been used in 1 publication

CCR9 Monoclonal specifically detects CCR9 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen CCR9
Applications Flow Cytometry
Classification Monoclonal
Clone 248621
Conjugate APC
Dilution Flow Cytometry 10 uL/10^6 cells
Gene Accession No. P51686
Gene Alias C-C chemokine receptor type 9, C-C CKR-9, CC-CKR-9, CCR9, CCR-9, CD199, CDw199, CDw199 antigen, chemokine (C-C motif) receptor 9, G protein-coupled receptor 28, GPR28, GPR-9-6G-protein coupled receptor 28
Gene Symbols CCR9
Host Species Mouse
Immunogen Human CCR9 transfectants Met1-Leu369 Accession # P51686
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 Tests
Primary or Secondary Primary
Gene ID (Entrez) 10803
Test Specificity Detects human CCR9 in flow cytometry.
Target Species Human
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.