Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human CCRL2/CRAM-A/B Alexa Fluor 750-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems FAB2350S100UG
This item is not returnable.
View return policy
Description
CCRL2/CRAM-A/B Monoclonal specifically detects CCRL2/CRAM-A/B in Human samples. It is validated for Flow Cytometry.Specifications
CCRL2/CRAM-A/B | |
Monoclonal | |
0.2 mg/mL | |
Flow Cytometry 0.25-1 ug/10^6 cells | |
NP_001124382 | |
CCRL2 | |
NS0 mouse myeloma cell line transfected with human CCRL2/CRAM-A/B Met1-Val356 (Val180Met) Accession # NP_001124382 | |
RUO | |
9034 | |
Human | |
IgG2b |
Flow Cytometry | |
152211 | |
Alexa Fluor 750 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
ACKR5, C-C chemokine receptor-like 2, CCR11, CCR6, CCRL2, chemokine (C-C motif) receptor-like 2, Chemokine receptor CCR11, Chemokine receptor X, CKRX, CKRXMGC116710, CRAM, CRAMA, CRAM-A, CRAMB, CRAM-B, FLJ55815, HCR, HCRMGC34104, Putative MCP-1 chemokine receptor | |
Mouse | |
100 μg | |
Primary | |
Detects human CCRL2/CRAM-A/B. Stains human HCR-transfected cells but not irrelevant transfectants. It detects both the CRAM-A and CRAM-B isoforms. | |
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction