Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human CCRL2/CRAM-A/B APC-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems FAB23501A
This item is not returnable.
View return policy
Description
CCRL2/CRAM-A/B Monoclonal specifically detects CCRL2/CRAM-A/B in Human samples. It is validated for Flow Cytometry.Specifications
CCRL2/CRAM-A/B | |
Monoclonal | |
APC | |
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
ACKR5, C-C chemokine receptor-like 2, CCR11, CCR6, CCRL2, chemokine (C-C motif) receptor-like 2, Chemokine receptor CCR11, Chemokine receptor X, CKRX, CKRXMGC116710, CRAM, CRAMA, CRAM-A, CRAMB, CRAM-B, FLJ55815, HCR, HCRMGC34104, Putative MCP-1 chemokine receptor | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
9034 | |
Human | |
Purified |
Flow Cytometry | |
152254 | |
Flow Cytometry 10 uL/10^6 cells | |
NP_001124382 | |
CCRL2 | |
NS0 mouse myeloma cell line transfected with human CRAM-A Met1-Val356 (Val180Met) Accession # NP_001124382 | |
100 Tests | |
Primary | |
Detects human CCRL2/CRAM-A/B. Stains human CCRL2-transfected cells but not irrelevant transfectants. Detects both the CRAM-A and CRAM-B isoforms. | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction