Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human CD-M6PR Lumenal Domain Biotinylated Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems BAF5320
This item is not returnable.
View return policy
Description
CD-M6PR Polyclonal specifically detects CD-M6PR in Human samples. It is validated for Western Blot.Specifications
CD-M6PR | |
Polyclonal | |
Biotin | |
000 Man6PR, 46 kDa mannose 6-phosphate receptor, 46-kDa mannose 6-phosphate receptor, cation-dependent mannose-6-phosphate receptor, CD Man-6-P receptor, CDM6PR, CD-MPR, FLJ32994, M6PR, mannose-6-phosphate receptor (cation dependent), MPR 46, MPR46, MPRD, Mr 46, SMPR | |
Goat | |
Affinity Purified | |
RUO | |
4074 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Western Blot | |
LYOPH | |
Western Blot 0.1 ug/mL | |
M6PR | |
Mouse myeloma cell line NS0-derived recombinant human CD-M6PR Lumenal Domain | |
50 μg | |
Primary | |
Detects human CD-M6PR Lumenal Domain in Western blots. In Western blots, approximately 40% cross-reactivity with recombinant mouse CD-M6PR is observed. | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction