Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human CD109 Alexa Fluor 488-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FAB4385G100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB4385G100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB4385G100 Supplier R&D Systems Supplier No. FAB4385G100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

CD109 Monoclonal specifically detects CD109 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen CD109
Applications Flow Cytometry
Classification Monoclonal
Clone 496920
Concentration 0.2 mg/mL
Conjugate Alexa Fluor 488
Dilution Flow Cytometry 0.25-1 ug/10^6 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
Gene Accession No. Q6YHK3
Gene Alias 150 kDa TGF-beta-1-binding protein, activated T-cell marker CD109, C3 and PZP-like alpha-2-macroglobulin domain-containing protein 7, CD109, CD109 antigen, CD109 antigen (Gov platelet alloantigens), CD109 molecule, CPAMD7, CPAMD7r150, DKFZp762L1111, FLJ38569, FLJ41966, Gov platelet alloantigens, p180, Platelet-specific Gov antigen, RP11-525G3.1
Gene Symbols CD109
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human CD109 Val22-Ser1268 (Tyr703Ser & Thr1241Met) Accession # Q6YHK3
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 135228
Test Specificity Detects human CD109 in direct ELISAs.
Target Species Human
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.