Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human CD109 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB4385SP
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB4385SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB4385SP Supplier R&D Systems Supplier No. MAB4385SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

CD109 Monoclonal specifically detects CD109 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, CyTOF-ready.

Specifications

Antigen CD109
Applications Flow Cytometry, Immunohistochemistry, CyTOF
Classification Monoclonal
Clone 496920
Concentration LYOPH
Conjugate Unconjugated
Dilution Flow Cytometry 2.5 ug/10^6 cells, Immunohistochemistry 8-25 ug/mL, CyTOF-ready
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q6YHK3
Gene Alias 150 kDa TGF-beta-1-binding protein, activated T-cell marker CD109, C3 and PZP-like alpha-2-macroglobulin domain-containing protein 7, CD109, CD109 antigen, CD109 antigen (Gov platelet alloantigens), CD109 molecule, CPAMD7, CPAMD7r150, DKFZp762L1111, FLJ38569, FLJ41966, Gov platelet alloantigens, p180, Platelet-specific Gov antigen, RP11-525G3.1
Gene Symbols CD109
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human CD109 Val22-Ser1268 (Tyr703Ser & Thr1241Met) Accession # Q6YHK3
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 135228
Test Specificity Detects human CD109 in direct ELISAs.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.