Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human CD117/c-kit APC-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Mouse Monoclonal Antibody has been used in 1 publication

$221.50 - $456.00

Specifications

Antigen CD117/c-kit
Clone 47233
Dilution Flow Cytometry 10 uL/10^6 cells
Applications Flow Cytometry
Classification Monoclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
FAB332A025
GREENER_CHOICE
View Documents
R&D Systems
FAB332A025
25 Tests
Each for $221.50
Only null left
Add to Cart
 
FAB332A100
GREENER_CHOICE
View Documents
R&D Systems
FAB332A100
100 Tests
Each for $456.00
Only null left
Add to Cart
 
Description

Description

CD117/c-kit Monoclonal specifically detects CD117/c-kit in Human samples. It is validated for Flow Cytometry.
Specifications

Specifications

CD117/c-kit
Flow Cytometry 10 uL/10^6 cells
Monoclonal
Purified
RUO
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
CD117, CD117 antigen, ckit, C-Kit, EC 2.7.10, EC 2.7.10.1, KIT, PBT, piebald trait, Proto-oncogene c-Kit, proto-oncogene tyrosine-protein kinase Kit, SCF R, SCFR, SCFRmast/stem cell growth factor receptor, soluble KIT variant 1, Tyrosine-protein kinase Kit, v-kit Hardy-Zuckerman 4 feline sarcoma viral oncogene homolog, v-kit Hardy-Zuckerman 4 feline sarcoma viral oncogene-like protein
KIT
IgG1
Protein A or G purified from ascites
Detects human CD117/c-kit in ELISAs and Western blots. In sandwich immunoassays and Western blots, no cross-reactivity with recombinant mouse CD117 is observed.
47233
Flow Cytometry
APC
Mouse
Human
P10721
3815
S. frugiperda insect ovarian cell line Sf 21-derived recombinant human CD117/c-kit Gln26-Thr520 Accession # P10721
Primary
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.