Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human CD117/c-kit Biotinylated Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems BAF332
This item is not returnable.
View return policy
Description
CD117/c-kit Polyclonal specifically detects CD117/c-kit in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA Detection (Matched Antibody Pair).Specifications
CD117/c-kit | |
Polyclonal | |
Biotin | |
P10721 | |
KIT | |
S. frugiperda insect ovarian cell line Sf 21-derived recombinant human CD117/c-kit Gln26-Thr520 Accession # P10721 | |
50 μg | |
Primary | |
Detects human CD117/c-kit in ELISAs and Western blots. In sandwich ELISAs, less than 0.1% cross-reactivity with recombinant mouse CD117, recombinant human (rh) PDGF R alpha , and rhPDGF R beta is observed. | |
Human | |
IgG |
Western Blot, Flow Cytometry, ELISA | |
LYOPH | |
Western Blot 0.1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, ELISA Detection (Matched Antibody Pair) 0.1-0.4 ug/mL | |
CD117, CD117 antigen, ckit, C-Kit, EC 2.7.10, EC 2.7.10.1, KIT, PBT, piebald trait, Proto-oncogene c-Kit, proto-oncogene tyrosine-protein kinase Kit, SCF R, SCFR, SCFRmast/stem cell growth factor receptor, soluble KIT variant 1, Tyrosine-protein kinase Kit, v-kit Hardy-Zuckerman 4 feline sarcoma viral oncogene homolog, v-kit Hardy-Zuckerman 4 feline sarcoma viral oncogene-like protein | |
Goat | |
Affinity Purified | |
RUO | |
3815 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction