Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human CD11b/Integrin alpha M Alexa Fluor™ 488-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
$151.20 - $339.00
Specifications
Antigen | CD11b |
---|---|
Clone | ICRF44 |
Dilution | Flow Cytometry 5 uL/10^6 cells |
Applications | Flow Cytometry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
FAB1699G025
![]() |
R&D Systems
FAB1699G025 |
25 Tests |
Each for $151.20
|
|
|||||
FAB1699G100
![]() |
R&D Systems
FAB1699G100 |
100 Tests |
Each for $339.00
|
|
|||||
Description
CD11b Monoclonal specifically detects CD11b in Human samples. It is validated for Flow Cytometry.Specifications
CD11b | |
Flow Cytometry 5 uL/10^6 cells | |
Monoclonal | |
Purified | |
RUO | |
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
3684 | |
Human rheumatoid synovial cells and monocytes | |
Primary | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. |
ICRF44 | |
Flow Cytometry | |
Alexa Fluor 488 | |
Mouse | |
Human | |
antigen CD11b (p170), CD11 antigen-like family member B, CD11b, CD11b antigen, Cell surface glycoprotein MAC-1 subunit alpha, Complement component 3 receptor 3 subunit, CR-3 alpha chain, CR3A, CR3AMGC117044, Integrin alpha M, integrin alpha-M, integrin, alpha M (complement component 3 receptor 3 subunit), ITGAM, Leukocyte adhesion receptor MO1, MAC-1, MAC1A, macrophage antigen alpha polypeptide, MO1A, Neutrophil adherence receptor, neutrophil adherence receptor alpha-M subunit, SLEB6 | |
ITGAM | |
IgG1 | |
Protein A or G purified from hybridoma culture supernatant | |
Detects human Integrin alpha M/CD11b. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title