Learn More
Human CD11c PE-conjugated Antibody, R&D Systems™
GREENER_CHOICE
Mouse Monoclonal Antibody has been used in 1 publication
Supplier:  R&D Systems FAB1777P100
 View more versions of this product
Catalog No. FAB1777P100
Only null left
Add to Cart
This item is not returnable. View return policy
Description
Description
CD11c Monoclonal specifically detects CD11c in Human samples. It is validated for Flow Cytometry.
Specifications
Specifications
CD11c
Monoclonal
R-PE
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
ITGAX
Rheumatoid synovial fluid cells and human monocyte-derived fibronectin
100 Tests
Primary
Detects human CD11c in flow cytometry.
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
IgG1
Flow Cytometry
ICRF 3.9
Flow Cytometry 10 uL/10^6 cells
95, 95 alpha chain, 95, alpha subunit, CD11 antigen-like family member C, CD11c, CD11c antigen, CD11Cleu M5, alpha subunit, Integrin alpha X, integrin alpha-X, integrin, alpha X (antigen CD11C (p150), alpha polypeptide), integrin, alpha X (complement component 3 receptor 4 subunit), ITGAX, Leu M5, Leukocyte adhesion glycoprotein p150, Leukocyte adhesion receptor p150, leukocyte surface antigen p150, myeloid membrane antigen, alpha subunit, p150 95 integrin alpha chain, p150,95 alpha, SLEB6
Mouse
Protein A or G purified from hybridoma culture supernatant
RUO
3687
Human
Purified
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.