Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human CD133 PE-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems FAB11331P100
This item is not returnable.
View return policy
Description
CD133 Monoclonal specifically detects CD133 in Human samples. It is validated for Flow Cytometry.Specifications
CD133 | |
Monoclonal | |
R-PE | |
Supplied in a saline solution containing BSA and Sodium Azide. | |
AC133, AC133 Stargardt disease 4 (autosomal dominant), CD133, CD133 retinal 2, PROM1, prominin (mouse)-like 1, prominin 1, PROML1 | |
NS0 mouse myeloma cell line transfected with human CD133 Accession # O43490 | |
100 Tests | |
Primary | |
Detects human CD133. Stains human CD133 transfectants but not irrelevant transfectants in flow cytometry. | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
IgG2a |
Flow Cytometry | |
170411 | |
Flow Cytometry 10 uL/10^6 cells | |
O43490 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
8842 | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
RUO
Spot an opportunity for improvement?Share a Content Correction