Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human CD133 PE-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FAB11331PL
Change view
Click to view available options
Quantity:
25 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB11331PL 25 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB11331PL Supplier R&D Systems Supplier No. FAB11331P025
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

CD133 Monoclonal specifically detects CD133 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen CD133
Applications Flow Cytometry
Classification Monoclonal
Clone 170411
Conjugate R-PE
Dilution Flow Cytometry 10 uL/10^6 cells
Formulation Supplied in a saline solution containing BSA and Sodium Azide.
Gene Accession No. O43490
Gene Alias AC133, AC133 Stargardt disease 4 (autosomal dominant), CD133, CD133 retinal 2, PROM1, prominin (mouse)-like 1, prominin 1, PROML1
Host Species Mouse
Immunogen NS0 mouse myeloma cell line transfected with human CD133 Accession # O43490
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8842
Test Specificity Detects human CD133. Stains human CD133 transfectants but not irrelevant transfectants in flow cytometry.
Target Species Human
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.