Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human CD14 Alexa Fluor 594-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
$221.50 - $397.50
Specifications
Antigen | CD14 |
---|---|
Clone | 134620 |
Dilution | Flow Cytometry 5 uL/10^6 cells |
Applications | Flow Cytometry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
FAB3832T025
![]() |
R&D Systems
FAB3832T025 |
25 Tests |
Each for $221.50
|
|
|||||
FAB3832T100
![]() |
R&D Systems
FAB3832T100 |
100 Tests |
Each for $397.50
|
|
|||||
Description
CD14 Monoclonal specifically detects CD14 in Human samples. It is validated for Flow Cytometry.Specifications
CD14 | |
Flow Cytometry 5 uL/10^6 cells | |
Monoclonal | |
Purified | |
RUO | |
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
CD14, CD14 antigen, CD14 molecule, monocyte differentiation antigen CD14, Myeloid cell-specific leucine-rich glycoprotein | |
CD14 | |
IgG1 | |
Protein A or G purified from hybridoma culture supernatant | |
Detects human CD14 in direct ELISAs and Western blots. |
134620 | |
Flow Cytometry | |
Alexa Fluor 594 | |
Mouse | |
Human | |
P08571 | |
929 | |
Chinese hamster ovary cell line CHO-derived recombinant human CD14 Thr20-Cys352 Accession # P08571 | |
Primary | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title