Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human CD14 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF3831SP
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF3831SP 25 μg
AF3831100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. AF3831SP Supplier R&D Systems Supplier No. AF3831SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

CD14 Polyclonal antibody specifically detects CD14 in Human samples. It is validated for Western Blot, Western Blot, Immunocytochemistry

Specifications

Antigen CD14
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Conjugate Unconjugated
Dilution 0.5 μg/mL (Western Blot), 20 μg/mL (Simple Western), 5 to 25 μg/mL (Immunocytochemistry)
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS.
Gene Accession No. P08571
Gene Alias CD14 antigen, CD14 molecule, monocyte differentiation antigen CD14, Myeloid cell-specific leucine-rich glycoprotein
Host Species Goat
Immunogen Chinese Hamster Ovary cell line, CHO-derived human CD14 protein, Thr20-Cys352, Accession # P08571
Purification Method Antigen Affinity-purified
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 929
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 6 months, -20°C to -70°C under sterile conditions after reconstitution.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.