Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human CD14 Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 16 publications
$241.00 - $2070.00
Specifications
Antigen | CD14 |
---|---|
Clone | 134620 |
Dilution | Western Blot 2 ug/mL, Flow Cytometry 0.25 ug/10^6 cells, Immunohistochemistry 5-25 ug/mL, Neutralization 10 ug/mL, CyTOF-ready |
Applications | Western Blot, Flow Cytometry, Immunohistochemistry, Neutralization, CyTOF |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB3832SP
![]() |
R&D Systems
MAB3832SP |
25 μg |
Each for $241.00
|
|
|||||
MAB3832100
![]() |
R&D Systems
MAB3832100 |
100 μg |
Each for $573.00
|
|
|||||
MAB3832500
![]() |
R&D Systems
MAB3832500 |
500 μg |
Each for $2,070.00
|
|
|||||
Description
CD14 Monoclonal specifically detects CD14 in Human samples. It is validated for Flow Cytometry, Neutralization, CyTOF-ready.Specifications
CD14 | |
Western Blot 2 ug/mL, Flow Cytometry 0.25 ug/10^6 cells, Immunohistochemistry 5-25 ug/mL, Neutralization 10 ug/mL, CyTOF-ready | |
Monoclonal | |
Mouse | |
Human | |
P08571 | |
929 | |
Chinese hamster ovary cell line CHO-derived recombinant human CD14 Thr20-Cys352 Accession # P08571 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human CD14 in direct ELISAs and Western blots. |
134620 | |
Western Blot, Flow Cytometry, Immunohistochemistry, Neutralization, CyTOF | |
Unconjugated | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
CD14, CD14 antigen, CD14 molecule, monocyte differentiation antigen CD14, Myeloid cell-specific leucine-rich glycoprotein | |
CD14 | |
IgG1 | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title