Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human CD14 Antibody, R&D Systems™

Mouse Monoclonal Antibody
$165.01 - $437.75
Specifications
| Antigen | CD14 |
|---|---|
| Clone | 50040R |
| Dilution | ELISA Capture (Matched Antibody Pair) 2-8 μg;/mL |
| Applications | ELISA |
| Classification | Monoclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB3833RSP
![]() |
R&D Systems
MAB3833RSP |
25 μg |
Each for $165.01
|
|
|||||
MAB3833R100
![]() |
R&D Systems
MAB3833R100 |
100 μg |
Each for $437.75
|
|
|||||
Description
CD14 Monoclonal specifically detects CD14 in Human samples. It is validated for ELISA Capture (Matched Antibody Pair).Specifications
| CD14 | |
| ELISA Capture (Matched Antibody Pair) 2-8 μg;/mL | |
| Monoclonal | |
| Supernatant | |
| Human | |
| P08571 | |
| 929.0 | |
| IgG1 | |
| Protein A or G purified from cell culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. |
| 50040R | |
| ELISA | |
| Unconjugated | |
| Mouse | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| CD14, CD14 antigen, CD14 molecule, monocyte differentiation antigen CD14, Myeloid cell-specific leucine-rich glycoprotein | |
| Chinese hamster ovary cell line CHO-derived recombinant human CD14, Thr20-Cys352, Accession No. P08571 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human CD14 in direct ELISAs. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title