Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human CD14 Biotinylated Antibody, R&D Systems™

Sheep Polyclonal Antibody has been used in 4 publications
Supplier: R&D Systems BAF383
This item is not returnable.
View return policy
Description
CD14 Polyclonal specifically detects CD14 in Human samples. It is validated for Western Blot, Immunohistochemistry, ELISA Detection (Matched Antibody Pair).Specifications
CD14 | |
Polyclonal | |
Biotin | |
P08571 | |
CD14 | |
Chinese hamster ovary cell line CHO-derived recombinant human CD14 Thr20-Cys352 Accession # P08571 | |
50 μg | |
Primary | |
Detects human CD14 in ELISAs and Western blots. In sandwich ELISAs, less than 0.2% cross-reactivity with recombinant mouse CD14 and Lipopolysaccharide is observed. | |
Human | |
IgG |
Western Blot, Immunohistochemistry, ELISA | |
LYOPH | |
Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL, ELISA Detection (Matched Antibody Pair) 0.1-0.4 ug/mL | |
CD14, CD14 antigen, CD14 molecule, monocyte differentiation antigen CD14, Myeloid cell-specific leucine-rich glycoprotein | |
Sheep | |
Affinity Purified | |
RUO | |
929 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction