Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Human CD14 Quantikine QuicKit ELISA

Catalog No. qk383
Click to view available options
Quantity:
1 Kit
This item is not returnable. View return policy
This item is not returnable. View return policy

From the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

  • A one step, 80-minute solid phase ELISA designed to measure human CD14 levels in cell culture supernates, serum, and plasma.
  • It contains CHO cell-expressed recombinant human CD14 and antibodies raised against the recombinant protein.
  • Results obtained for naturally occurring human CD14 showed linear curves that were parallel to the standard curves obtained using the recombinant QuicKit™ standards.
  • These results indicate that this kit can be used determine relative mass values for natural CD14.

Specifications

Product Type ELISA
Specificity Natural and Recombinant Human CD14
Gene ID (Entrez) 929
Host Species Human
Quantity 1 Kit
Storage Requirements Store the unopened product at 2 - 8 °C. Do not use past expiration date.
Synonym CD14, CD14 antigen, CD14 molecule, monocyte differentiation antigen CD14, Myeloid cell-specific leucine-rich glycoprotein
Target CD14
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.