Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human CD15/Lewis X Alexa Fluor 488-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems FAB7368G100UG
This item is not returnable.
View return policy
Description
CD15/Lewis X Monoclonal specifically detects CD15/Lewis X in Human samples. It is validated for Flow Cytometry.Specifications
CD15/Lewis X | |
Monoclonal | |
Alexa Fluor 488 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and sodium azide | |
FUT4 | |
Human granulocytes | |
100 μg | |
Detects human CD15/Lewis X in direct ELISAs. | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied. | |
IgM |
Flow Cytometry | |
ICRF29-2 | |
Flow Cytometry 0.25-1 ug/10^6 cells | |
CD 15, CD15, CD-15, Lewis X, Lewis-X, LeX, SSEA-1, stage-specific embryonic antigen 1 | |
Mouse | |
IgM-specific Affinity-purified from hybridoma culture supernatant | |
Primary | |
Human | |
Supernatant |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction