Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human CD15/Lewis X Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB7368SP
This item is not returnable.
View return policy
Description
CD15/Lewis X Monoclonal specifically detects CD15/Lewis X in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, CyTOF-ready.Specifications
CD15/Lewis X | |
Monoclonal | |
LYOPH | |
Flow Cytometry 2.5 ug/10^6 cells, Immunohistochemistry 5-25 ug/mL, CyTOF-ready | |
CD 15, CD15, CD-15, Lewis X, Lewis-X, LeX, SSEA-1, stage-specific embryonic antigen 1 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
2526 | |
Sterile PBS to a final concentration of 0.5 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgM |
Flow Cytometry, Immunohistochemistry, CyTOF | |
ICRF29-2 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
FUT4 | |
human granulocytes | |
25 μg | |
Primary | |
Detects human CD15/Lewis X in direct ELISAs. | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction