Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human CD155/PVR Alexa Fluor 488-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FA25301G100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FA25301G100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FA25301G100 Supplier R&D Systems Supplier No. FAB25301G100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

CD155/PVR Monoclonal specifically detects CD155/PVR in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen CD155/PVR
Applications Flow Cytometry
Classification Monoclonal
Clone 300907
Concentration 0.2 mg/mL
Conjugate Alexa Fluor 488
Dilution Flow Cytometry 0.25-1 ug/10^6 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
Gene Accession No. AAH15542
Gene Alias CD155, CD155 antigen, HVED, Necl-5, NECL5, nectin-like 5, Nectin-like protein 5, poliovirus receptor, PVR, PVS, PVSFLJ25946, Tage4
Gene Symbols PVR
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human CD155/PVR Gly27-Asn343 Accession # AAH15542
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5817
Test Specificity Detects human CD155/PVR in direct ELISAs and Western blots.
Target Species Human
Content And Storage Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Isotype IgG1
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.