Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human CD155/PVR Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB2530SP
This item is not returnable.
View return policy
Description
CD155/PVR Monoclonal specifically detects CD155/PVR in Human samples. It is validated for Western Blot.Specifications
CD155/PVR | |
Monoclonal | |
LYOPH | |
Western Blot 1 ug/mL | |
ABM85069 | |
PVR | |
Mouse myeloma cell line NS0-derived recombinant human CD155/PVR Gly27-Asn343 Accession # ABM85069 | |
25 μg | |
Primary | |
Detects human CD155/PVR in direct ELISAs and Western blots. | |
Human | |
Purified |
Western Blot | |
300903 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
CD155, CD155 antigen, HVED, Necl-5, NECL5, nectin-like 5, Nectin-like protein 5, poliovirus receptor, PVR, PVS, PVSFLJ25946, Tage4 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
5817 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction