Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human CD161 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB7448
This item is not returnable.
View return policy
Description
CD161 Monoclonal specifically detects CD161 in Human samples. It is validated for Flow Cytometry, CyTOF-ready.Specifications
CD161 | |
Monoclonal | |
LYOPH | |
Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready | |
Q12918 | |
KLRB1 | |
BaF3 mouse pro-B cell line transfected with human CD161 Accession # Q12918 | |
100 μg | |
Primary | |
Detects human CD161 in direct ELISAs. | |
Human | |
Purified |
Flow Cytometry, CyTOF | |
702228 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
CD161, CD161 antigen, CLEC5B, CLEC5BC-type lectin domain family 5 member B, HNKR-P1a, killer cell lectin-like receptor subfamily B member 1, killer cell lectin-like receptor subfamily B, member 1, KLRB1, Ly59, Natural killer cell surface protein P1A, NKR, NKRP1, NKR-P1, NKRP1A, NKR-P1AMGC138614, NKRP1ANKR | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
3820 | |
Sterile PBS to a final concentration of 0.5 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction