Learn More
Human CD163 Alexa Fluor™ 405-conjugated Antibody, R&D Systems™
GREENER_CHOICE
Mouse Monoclonal Antibody
Supplier:  R&D Systems FAB16072V100UG
Catalog No. FAB16072V10
Only null left
Add to Cart
This item is not returnable. View return policy
Description
Description
CD163 Monoclonal specifically detects CD163 in Human samples. It is validated for Flow Cytometry.
Specifications
Specifications
CD163
Monoclonal
Alexa Fluor 405
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide.
Mouse
Protein A or G purified from hybridoma culture supernatant
Primary
Detects human CD163 in Flow Cytometry.
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
IgG1
Flow Cytometry
R20
Flow Cytometry 0.25-1 ug/10^6 cells
CD_antigen: CD163, CD163, CD163 molecule, GHI/61, HbSR, Hemoglobin scavenger receptor, M130, macrophage-associated antigen, MM130, RM3/1, SCARI1, scavenger receptor cysteine-rich type 1 protein M130, sCD163, Soluble CD163
Balb/c mice immunized with Synovial cells from Rheumatoid Arthritis patients
100 μg
9332
Human
Supernatant
Product Suggestions
Product Suggestions
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.