Learn More
Human CD163 Alexa Fluor™ 488-conjugated Antibody, R&D Systems™
GREENER_CHOICE
Mouse Monoclonal Antibody has been used in 2 publications
Supplier:  R&D Systems FAB1607G025
 View more versions of this product
Catalog No. FAB1607G025
Only null left
Add to Cart
This item is not returnable. View return policy
Description
Description
CD163 Monoclonal specifically detects CD163 in Human samples. It is validated for Flow Cytometry.
Specifications
Specifications
CD163
Monoclonal
Alexa Fluor 488
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
CD_antigen: CD163, CD163, CD163 molecule, GHI/61, HbSR, Hemoglobin scavenger receptor, M130, macrophage-associated antigen, MM130, RM3/1, SCARI1, scavenger receptor cysteine-rich type 1 protein M130, sCD163, Soluble CD163
Mouse
Protein A or G purified from hybridoma culture supernatant
RUO
9332
Human
Purified
Flow Cytometry
215927
Flow Cytometry 5 uL/10^6 cells
Q86VB7
CD163
Mouse myeloma cell line NS0-derived recombinant human CD163 Gly46-Ser1050 Accession # Q86VB7
25 Tests
Primary
Detects human CD163 in direct ELISAs and Western blots.
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
IgG1
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.