Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human CD163 Antibody, R&D Systems™

Mouse Monoclonal Antibody
$200.50 - $2070.00
Specifications
Antigen | CD163 |
---|---|
Clone | 215930 |
Concentration | LYOPH |
Dilution | ELISA Capture (Matched Antibody Pair) 2-8 ug/mL |
Applications | ELISA |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB16071SP
![]() |
R&D Systems
MAB16071SP |
25 μg |
Each for $200.50
|
|
|||||
MAB16071100
![]() |
R&D Systems
MAB16071100 |
100 μg |
Each for $561.50
|
|
|||||
MAB16071500
![]() |
R&D Systems
MAB16071500 |
500 μg |
Each for $2,070.00
|
|
|||||
Description
CD163 Monoclonal specifically detects CD163 in Human samples. It is validated for ELISA Capture (Matched Antibody Pair).Specifications
CD163 | |
LYOPH | |
ELISA | |
Unconjugated | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
CD_antigen: CD163, CD163, CD163 molecule, GHI/61, HbSR, Hemoglobin scavenger receptor, M130, macrophage-associated antigen, MM130, RM3/1, SCARI1, scavenger receptor cysteine-rich type 1 protein M130, sCD163, Soluble CD163 | |
CD163 | |
IgG1 | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
215930 | |
ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
Monoclonal | |
Mouse | |
Human | |
CAA80543 | |
9332 | |
Mouse myeloma cell line NS0-derived recombinant human CD163 Gly41-Ser1045 Accession # CAA80543 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human CD163 in direct ELISAs. In sandwich immunoassays, no cross-reactivity or interference with recombinant mouse (rm) CD163, rmCD6, or recombinant human CD6 is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title