Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human CD164 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB5790
This item is not returnable.
View return policy
Description
CD164 Monoclonal specifically detects CD164 in Human samples. It is validated for Flow Cytometry.Specifications
CD164 | |
Monoclonal | |
LYOPH | |
Flow Cytometry 0.25 ug/10^6 cells | |
Q04900 | |
CD164 | |
NS0 mouse myeloma cell line transfected with human CD14 Asp24-Leu197 Accession # Q04900 | |
100 μg | |
Primary | |
Detects human CD164 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant mouse CD164 is observed. | |
Human | |
Purified |
Flow Cytometry | |
502021 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in Tris and NaCl with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
CD164, CD164 antigen, CD164 molecule, sialomucin, Endolyn, MGC-24, MGC-24v, MUC-24, Multi-glycosylated core protein 24, sialomucin | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
8763 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG3 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction