Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human CD164 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB5790SP
This item is not returnable.
View return policy
Description
CD164 Monoclonal specifically detects CD164 in Human samples. It is validated for Flow Cytometry.Specifications
| CD164 | |
| Monoclonal | |
| LYOPH | |
| Flow Cytometry 0.25 ug/10^6 cells | |
| Q04900 | |
| CD164 | |
| NS0 mouse myeloma cell line transfected with human CD14 Asp24-Leu197 Accession # Q04900 | |
| 25 μg | |
| Primary | |
| Detects human CD164 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant mouse CD164 is observed. | |
| Human | |
| Purified |
| Flow Cytometry | |
| 502021 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in Tris and NaCl with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CD164, CD164 antigen, CD164 molecule, sialomucin, Endolyn, MGC-24, MGC-24v, MUC-24, Multi-glycosylated core protein 24, sialomucin | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 8763 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG3 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction