Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human CD177 ELISA Kit

Catalog No. p-7219741
Encompass_Preferred
Click to view available options
Quantity:
5 x 96 Tests
96 Tests

Catalog No. EH80RBX5

Supplier: Invitrogen™ EH80RBX5

Only null left
Add to Cart
Add to Cart

ELISA

Human CD177 quantitates human CD177 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human CD177.

CD177 (NB1/HNA-2a and PRV-1 form) is a GPI-anchored glycoprotein present mainly on neutrophils. Its plasma membrane expression is increased during pregnancy and and inflammation or after G-CSF application. Ligand of CD177 has been identified as CD31 (PECAM-1). CD177 participates in neutrophil transmigration and seems to be also a pro-proliferative molecule. The antibodies against CD177 can be involved in neonatal alloimmune neutropenia (NAN).
TRUSTED_SUSTAINABILITY

Specifications

Accession Number 1190003K14Rik; Cd177; CD177 antigen; CD177 molecule; cell surface receptor; HNA2A; HNA-2 A; Human neutrophil alloantigen 2 A; NB1; NB1 glycoprotein; NB1 GP; Pdp3; Polycythemia rubra vera protein 1; PRV1; PRV-1; RGD1562941; UNQ595/PRO1181
Assay Range 0.21 to 50 ng/mL
Assay Sensitivity 0.21 ng/mL
Conjugate HRP
Product Type ELISA
Sample Type Plasma, Serum, Supernatant
For Use With (Equipment) Colorimetric Microplate Reader
Gene ID (Entrez) 57126
Gene Symbol CD177
Interassay CV <12%
Intraassay CV <10
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye HRP
Quantity 5 x 96 Tests
Regulatory Status RUO
Sample Volume Plasma 50 μL, Serum 50 μL, Supernatant 50 μL
Storage Requirements 2°C to 8°C
Target CD177
Test Time 1 hrs 20 mins
Total Assay Time 4 hrs 45 mins
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.