Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human CD19 Alexa Fluor 594-conjugated Antibody, R&D Systems™

Catalog No. FAB4867T025
Change view
Click to view available options
Quantity:
100 Tests
25 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB4867T025 25 Tests
FAB4867T100 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. FAB4867T025 Supplier R&D Systems Supplier No. FAB4867T025
This item is not returnable. View return policy

Mouse Monoclonal Antibody

CD19 Monoclonal specifically detects CD19 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen CD19
Applications Flow Cytometry
Classification Monoclonal
Clone 4G7-2E3
Conjugate Alexa Fluor 594
Dilution Flow Cytometry 5 uL/10^6 cells
Formulation Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
Gene Alias B4, B-lymphocyte antigen CD19, B-lymphocyte surface antigen B4, CD19, CD19 antigen, CD19 molecule, CVID3, Differentiation antigen CD19, Leu-12, MGC12802, T-cell surface antigen Leu-12
Gene Symbols CD19
Host Species Mouse
Immunogen Human CLL cells
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 930
Test Specificity Detects human CD19.
Target Species Human
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Form Supernatant
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.