Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human CD1a PE-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems FAB7076P
This item is not returnable.
View return policy
Description
CD1a Monoclonal specifically detects CD1a in Human samples. It is validated for Flow Cytometry.Specifications
CD1a | |
Monoclonal | |
R-PE | |
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
CD1, CD1a, CD1a antigen, CD1A antigen, a polypeptide, CD1a molecule, cluster of differentiation 1 A, cortical thymocyte antigen CD1A, differentiation antigen CD1-alpha-3, epidermal dendritic cell marker CD1a, FCB6, HTA1, hTa1 thymocyte antigen, R4, T6, T-cell surface antigen T6/Leu-6, T-cell surface glycoprotein CD1a | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
909 | |
Human | |
Purified |
Flow Cytometry | |
703217 | |
Flow Cytometry 10 uL/10^6 cells | |
P06126 | |
CD1A | |
Mouse myeloma cell line NS0-derived recombinant human CD1a Asp19-Val300 (predicted) Accession # P06126 | |
100 Tests | |
Primary | |
Detects human CD1a in direct ELISAs. In direct ELISAs, less than 5% cross-reactivity with recombinant human (rh) CD1d is observed and no cross-reactivity with with rhCD1b or rhCD1e is observed. | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction