Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human CD1b Alexa Fluor 405-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems FAB7446V100UG
This item is not returnable.
View return policy
Description
CD1b Monoclonal specifically detects CD1b in Human samples. It is validated for Flow Cytometry.Specifications
CD1b | |
Monoclonal | |
0.2 mg/mL | |
Flow Cytometry 0.25-1 ug/10^6 cells | |
P29016 | |
CD1B | |
Mouse myeloma cell line NS0-derived recombinant human CD1b Ser18-Ser303 Accession # P29016 | |
RUO | |
910 | |
Human | |
IgG2b |
Flow Cytometry | |
737249 | |
Alexa Fluor 405 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
CD1, CD1A, CD1b, CD1b antigen, CD1B antigen, b polypeptide, CD1b molecule, cortical thymocyte antigen CD1B, differentiation antigen CD1-alpha-3, MGC125990, MGC125991, R1, T-cell surface glycoprotein CD1b | |
Mouse | |
100 μg | |
Primary | |
Detects human CD1b in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human (rh) CD1a, rhCD1c, rhCD1d, rhCD1e, recombinant mouse (rm) CD1d1, or rmCD1d2 is observed. | |
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction