Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human CD1b Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB7446SP
This item is not returnable.
View return policy
Description
CD1b Monoclonal specifically detects CD1b in Human samples. It is validated for Flow Cytometry, CyTOF-ready.Specifications
CD1b | |
Monoclonal | |
LYOPH | |
Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready | |
P29016 | |
CD1B | |
Mouse myeloma cell line NS0-derived recombinant human CD1b Ser18-Ser303 Accession # P29016 | |
25 μg | |
Primary | |
Detects human CD1b in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human (rh) CD1a, rhCD1c, rhCD1d, rhCD1e, recombinant mouse (rm) CD1d1, or rmCD1d2 is observed. | |
Human | |
Purified |
Flow Cytometry, CyTOF | |
737249 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
CD1, CD1A, CD1b, CD1b antigen, CD1B antigen, b polypeptide, CD1b molecule, cortical thymocyte antigen CD1B, differentiation antigen CD1-alpha-3, MGC125990, MGC125991, R1, T-cell surface glycoprotein CD1b | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
910 | |
Sterile PBS to a final concentration of 0.5 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction