Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human CD1c/BDCA-1 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF5910
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF5910 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF5910 Supplier R&D Systems Supplier No. AF5910
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

CD1c/BDCA-1 Polyclonal specifically detects CD1c/BDCA-1 in Human samples. It is validated for Western Blot.

Specifications

Antigen CD1c/BDCA-1
Applications Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL
Gene Accession No. P29017
Gene Alias BDCA1, BDCA-1, c polypeptide, CD1, CD1A, CD1c, CD1c antigen, CD1c molecule, cortical thymocyte antigen CD1C, differentiation antigen CD1-alpha-3, R7, T-cell surface glycoprotein CD1c
Gene Symbols CD1C
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant human CD1c/BDCA-1 Ala19-Met302 (Phe300Ser) Accession # P29017
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 911
Test Specificity Detects human CD1c in direct ELISAs and Western blots. In direct ELISAs, less than 1% cross-reactivity with recombinant human (rh) CD1a, rhCD1b, rhCD1d, and rhCD1e is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.