Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human CD1e Alexa Fluor 700-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. IC7330N100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
IC7330N100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. IC7330N100 Supplier R&D Systems Supplier No. IC7330N100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

CD1e Monoclonal specifically detects CD1e in Human samples. It is validated for Intracellular Staining by Flow Cytometry.

Specifications

Antigen CD1e
Applications Flow Cytometry
Classification Monoclonal
Clone 704407
Concentration 0.2 mg/mL
Conjugate Alexa Fluor 700
Dilution Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
Gene Accession No. P15812
Gene Alias CD1-alpha-3, CD1e, CD1e molecule, CDIEL, e polypeptide, FLJ17609, membrane-associated, R2G1, thymocyte antigen CD1E
Gene Symbols CD1E
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human CD1e Met1-Tyr304 Accession # P15812
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 913
Test Specificity Detects human CD1e in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human (rh) CD1a, rhCD1b, rhCD1c, rhCD1d, or recombinant rat beta 2-Microglobulin is observed.
Target Species Human
Content And Storage Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Isotype IgG1
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.