Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human CD2 Antibody, R&D Systems™

Goat Polyclonal Antibody
$200.50 - $538.00
Specifications
Antigen | CD2 |
---|---|
Concentration | LYOPH |
Dilution | Western Blot 0.5 ug/mL, Simple Western 10 ug/mL, Flow Cytometry 0.25 ug/10^6 cells, Immunohistochemistry 3-15 ug/mL, CyTOF-ready |
Applications | Western Blot, Flow Cytometry, Immunohistochemistry, CyTOF |
Classification | Polyclonal |
Description
CD2 Polyclonal specifically detects CD2 in Human samples. It is validated for Western Blot, Simple Western, Flow Cytometry, CyTOF-ready.Specifications
CD2 | |
Western Blot 0.5 ug/mL, Simple Western 10 ug/mL, Flow Cytometry 0.25 ug/10^6 cells, Immunohistochemistry 3-15 ug/mL, CyTOF-ready | |
Polyclonal | |
Goat | |
Human | |
CD2, CD2 antigen, CD2 antigen (p50), sheep red blood cell receptor, CD2 molecule, Erythrocyte receptor, FLJ46032, LFA-2, LFA-3 receptor, lymphocyte-function antigen-2, Rosette receptor, SRBC, T11, T-cell surface antigen CD2, T-cell surface antigen T11/Leu-5 | |
CD2 | |
IgG | |
Affinity Purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
LYOPH | |
Western Blot, Flow Cytometry, Immunohistochemistry, CyTOF | |
Unconjugated | |
RUO | |
P06729 | |
914 | |
Mouse myeloma cell line NS0-derived recombinant human CD2 Lys25-Asp209 Accession # P06729 | |
Primary | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Detects human CD2 in direct ELISAs and Western blots. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title