Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human CD2 Antibody, R&D Systems™

Mouse Monoclonal Antibody
$200.50 - $538.00
Specifications
Antigen | CD2 |
---|---|
Clone | 299808 |
Dilution | Western Blot 2μg/mL, Agonist Activity 5μg/mL |
Applications | ChIP Assay, Western Blot |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB18562SP
![]() |
R&D Systems
MAB18562SP |
25 μg |
Each for $200.50
|
|
|||||
MAB18562100
![]() |
R&D Systems
MAB18562100 |
100 μg |
Each for $538.00
|
|
|||||
Description
CD2 Monoclonal specifically detects CD2 in Human samples. It is validated for Western Blot, Flow Cytometry.Specifications
CD2 | |
Western Blot 2μg/mL, Agonist Activity 5μg/mL | |
Monoclonal | |
Purified | |
Human | |
P06729 | |
914 | |
IgG1 | |
Protein A or G purified from ascites | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
299808 | |
ChIP Assay, Western Blot | |
Unconjugated | |
Mouse | |
Lyophilized from a 0.2μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2μm filtered solution in PBS. | |
CD2, CD2 antigen, CD2 antigen (p50), sheep red blood cell receptor, CD2 molecule, Erythrocyte receptor, FLJ46032, LFA-2, LFA-3 receptor, lymphocyte-function antigen-2, Rosette receptor, SRBC, T11, T-cell surface antigen CD2, T-cell surface antigen T11/Leu-5 | |
Mouse myeloma cell line NS0-derived recombinant human CD2 Lys25-Asp209 Accession # P06729 | |
Primary | |
Reconstitute at 0.5mg/mL in sterile PBS. | |
Detects human CD2 in direct ELISAs. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title