Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human CD20 Alexa Fluor 405-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
$214.00
Specifications
Antigen | CD20 |
---|---|
Clone | 396444 |
Dilution | Flow Cytometry 5 uL/10^6 cells |
Applications | Flow Cytometry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
FAB4225V025
![]() |
R&D Systems
FAB4225V025 |
25 Tests |
Each of 1 for $214.00
|
|
|||||
Description
CD20 Monoclonal specifically detects CD20 in Human samples. It is validated for Flow Cytometry.Specifications
CD20 | |
Flow Cytometry 5 uL/10^6 cells | |
Monoclonal | |
Supernatant | |
RUO | |
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
B1, B-lymphocyte antigen CD20, B-lymphocyte cell-surface antigen B1, B-lymphocyte surface antigen B1, Bp35, Bp35MGC3969, CD20, CD20 antigen, CD20 receptor, CD20S7, CVID5, LEU-16, Leukocyte surface antigen Leu-16, Ly-44, Membrane-spanning 4-domains subfamily A member 1, membrane-spanning 4-domains, subfamily A, member 1, MS4A1, MS4A2, S7 | |
MS4A1 | |
Primary | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. |
396444 | |
Flow Cytometry | |
Alexa Fluor 405 | |
Mouse | |
Human | |
P11836 | |
931 | |
IgG1 | |
Protein A or G purified from hybridoma culture supernatant | |
Detects human MS4A1/CD20. Stains human MS4A1/CD20 transfectants but not irrelevant transfectants in flow cytometry. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title