Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human CD20 Alexa Fluor 488-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems FAB4225G100UG
This item is not returnable.
View return policy
Description
CD20 Monoclonal specifically detects CD20 in Human samples. It is validated for Flow Cytometry.Specifications
| CD20 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Flow Cytometry 0.25-1 ug/10^6 cells | |
| P11836 | |
| MS4A1 | |
| NS0 mouse myeloma cell line transfected with human MS4A1/CD20 Met1-Pro297 Accession # P11836 | |
| 100 μg | |
| Primary | |
| Detects human MS4A1/CD20. Stains human MS4A1/CD20 transfectants but not irrelevant transfectants. | |
| Human | |
| IgG1 |
| Flow Cytometry | |
| 396444 | |
| Alexa Fluor 488 | |
| Supplied in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| B1, B-lymphocyte antigen CD20, B-lymphocyte cell-surface antigen B1, B-lymphocyte surface antigen B1, Bp35, Bp35MGC3969, CD20, CD20 antigen, CD20 receptor, CD20S7, CVID5, LEU-16, Leukocyte surface antigen Leu-16, Ly-44, Membrane-spanning 4-domains subfamily A member 1, membrane-spanning 4-domains, subfamily A, member 1, MS4A1, MS4A2, S7 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 931 | |
| Reconstitution default not known | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 °C as supplied. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction