Learn More
Human CD20 (Research Grade Rituximab Biosimilar) Alexa Fluor 350-conjugated Antibody, R&D Systems™
GREENER_CHOICE
Human Monoclonal Antibody
Supplier:  R&D Systems FAB9575U100UG
Catalog No. FAB9575U100
Only null left
Add to Cart
This item is not returnable. View return policy
Description
Description
CD20 Monoclonal specifically detects CD20 in Human samples. It is validated for Flow Cytometry.
Specifications
Specifications
CD20
Monoclonal
Alexa Fluor 350
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide.
Human
Protein A or G purified from cell culture supernatant
Primary
Detects Human CD20 based on Rituximab therapeutic antibody. This non-therapeutic antibody uses the same variable region sequence as the therapeutic antibody Rituximab. This product is for research use only.
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
IgG1
Flow Cytometry
Hu2
Flow Cytometry 0.25-1 ug/10^6 cells
B1, B-lymphocyte antigen CD20, B-lymphocyte cell-surface antigen B1, B-lymphocyte surface antigen B1, Bp35, Bp35MGC3969, CD20, CD20 antigen, CD20 receptor, CD20S7, CVID5, LEU-16, Leukocyte surface antigen Leu-16, Ly-44, Membrane-spanning 4-domains subfamily A member 1, membrane-spanning 4-domains, subfamily A, member 1, MS4A1, MS4A2, S7
Human CD20
100 μg
931
Human
Supernatant
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.