Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human CD200 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB27243SP
Change view
Click to view available options
Quantity:
100 μg
25 μg
500 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB27243SP 25 μg
MAB27243100 100 μg
MAB27243500 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. MAB27243SP Supplier R&D Systems Supplier No. MAB27243SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

CD200/OX2 Monoclonal specifically detects CD200/OX2 in Human samples. It is validated for Blockade of Receptor-ligand Interaction, ELISA Capture (Matched Antibody Pair).

Specifications

Antigen CD200/OX2
Applications Blockade of Receptor-ligand Interaction, ELISA
Classification Monoclonal
Clone 325531
Conjugate Unconjugated
Dilution Blockade of Receptor-ligand Interaction, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. P41217.3
Gene Alias antigen identified by monoclonal MRC OX-2, CD200, CD200 antigenMOX1, CD200 molecule, MOX1, MOX2, MOX2MRC, MRC, MRC OX-2 antigen, OX-2, OX-2 membrane glycoprotein
Gene Symbols CD200
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human CD200 Gln31-Gly232 Accession # P41217.3
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4345
Test Specificity Detects human CD200 in ELISAs. In sandwich immunoassays, no cross-reactivity or interference with recombinant mouse CD200 is observed.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.