Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human CD200 Biotinylated Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems BAF2724
This item is not returnable.
View return policy
Description
CD200/OX2 Polyclonal specifically detects CD200/OX2 in Human samples. It is validated for Western Blot.Specifications
CD200/OX2 | |
Polyclonal | |
Biotin | |
P41217 | |
CD200 | |
Mouse myeloma cell line NS0-derived recombinant human CD200 Gln31-Gly232 Accession # P41217 | |
50 μg | |
Primary | |
Detects human CD200 in Western blots. In this format, approximately 10% cross-reactivity with recombinant mouse CD200 is observed. | |
Human | |
IgG |
Western Blot | |
LYOPH | |
Western Blot 0.1 ug/mL | |
antigen identified by monoclonal MRC OX-2, CD200, CD200 antigenMOX1, CD200 molecule, MOX1, MOX2, MOX2MRC, MRC, MRC OX-2 antigen, OX-2, OX-2 membrane glycoprotein | |
Goat | |
Affinity Purified | |
RUO | |
4345 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction