Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human CD21 Antibody, R&D Systems™

Sheep Polyclonal Antibody
Supplier: R&D Systems AF4909
This item is not returnable.
View return policy
Description
CD21 Polyclonal specifically detects CD21 in Human samples. It is validated for Western Blot, Flow Cytometry, CyTOF-ready.Specifications
CD21 | |
Polyclonal | |
Unconjugated | |
P20023 | |
CR2 | |
Chinese hamster ovary cell line CHO-derived recombinant human CD21 Ile21-Arg971 Accession # P20023 | |
100 μg | |
Primary | |
Detects human CD21 in direct ELISAs and Western blots. | |
Human | |
IgG |
Immunohistochemistry, Flow Cytometry, CyTOF | |
LYOPH | |
Western Blot 0.1 μg/mL, Flow Cytometry 2.5 μg/106 cells, Immunohistochemistry 3 to 15 μg/mL, CyTOF-ready | |
C3DR, C3DRSLEB9, CD21, CD21 antigen, Complement C3d receptor, complement component (3d/Epstein Barr virus) receptor 2, complement receptor type 2, Cr2, EBV receptor, Epstein-Barr virus receptor | |
Sheep | |
Antigen Affinity-purified | |
RUO | |
1380 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction