Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human CD229/SLAMF3 Antibody, R&D Systems™

Goat Polyclonal Antibody
$200.50 - $549.50
Specifications
Antigen | CD229/SLAMF3/Lymphocyte Antigen 9 |
---|---|
Concentration | LYOPH |
Dilution | Western Blot 0.1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready |
Applications | Western Blot, Flow Cytometry, CyTOF |
Classification | Polyclonal |
Description
CD229/SLAMF3/Lymphocyte Antigen 9 Polyclonal specifically detects CD229/SLAMF3/Lymphocyte Antigen 9 in Human samples. It is validated for Western Blot, Flow Cytometry, CyTOF-ready.Specifications
CD229/SLAMF3/Lymphocyte Antigen 9 | |
Western Blot 0.1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready | |
Polyclonal | |
Goat | |
Human | |
CD229, CD229 antigen, cell-surface molecule Ly-9, hly9, Ly9, lymphocyte antigen 9Cell surface molecule Ly-9, mLY9, SLAMF3, T-lymphocyte surface antigen Ly-9 | |
LY9 | |
IgG | |
Affinity Purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
LYOPH | |
Western Blot, Flow Cytometry, CyTOF | |
Unconjugated | |
RUO | |
Q9HBG7 | |
4063 | |
Mouse myeloma cell line NS0-derived recombinant human CD229/SLAMF3 Lys48-Lys454 Accession # Q9HBG7 | |
Primary | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Detects human CD229/SLAMF3 in direct ELISAs and Western blots. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title