Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human CD23/Fc epsilon RII Alexa Fluor™ 350-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FAB123U100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB123U100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB123U100 Supplier R&D Systems Supplier No. FAB123U100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

CD23/Fc epsilon RII Monoclonal specifically detects CD23/Fc epsilon RII in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen CD23/Fc epsilon RII
Applications Flow Cytometry
Classification Monoclonal
Clone 138628
Concentration 0.2 mg/mL
Conjugate Alexa Fluor 350
Dilution Flow Cytometry 0.25-1 ug/10^6 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
Gene Accession No. P06734
Gene Alias BLAST-2, CD23, CD23A, CD23CD23 antigen, CLEC4J, CLEC4JC-type lectin domain family 4 member J, C-type lectin domain family 4, member J, Fc epsilon RII, Fc fragment of IgE, low affinity II, receptor for (CD23), FCE2Fc fragment of IgE, low affinity II, receptor for (CD23A), fc-epsilon-RII, FCER2, Fcer2a, FceRII, IGEBF, Immunoglobulin E-binding factor, low affinity immunoglobulin epsilon Fc receptor, Ly-42, Lymphocyte IgE receptor
Gene Symbols FCER2
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human CD23/Fc epsilon RII Met150-Ser321 Accession # P06734
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2208
Test Specificity Detects human CD23/Fc epsilon RII in direct ELISAs and Western blots.
Target Species Human
Content And Storage Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Isotype IgG1
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.